tert-Butyl-3-cyano-4-oxopyrrolidine-1-carboxylate ≥97.0%

Synonyms: 3-Cyano-4-oxopyrrolidine-1-carboxylic acid tert-Butyl ester, tert-Butyl 3-cyano-4-oxopyrrolidine-1-carboxylate, 1-Boc-3-cyano-4-pyrrolidone, tert-Butyl 3-Cyano-4-oxopyrrolidine-1-carboxylate, 1-tert-Butoxycarbonyl-3-cyano-4-pyrrolidone, 1-Boc-3-cyano-4-pyrrolidinone, 1-tert-Butoxycarbonyl-3-cyano-4-pyrrolidinone

02216-1 02216-2 02216-3
103534-656EA 23.08 USD
103534-656 103534-658 103534-660
tert-Butyl-3-cyano-4-oxopyrrolidine-1-carboxylate ≥97.0%
tert-Butyl-3-cyano-4-oxopyrrolidine-1-carboxylate
Formula: C₁₀H₁₄N₂O₃
MW: 210.23 g/mol
MDL Number: MFCD01318456
CAS Number: 175463-32-8

Order Now

Specification Test Results

Purety 97.0%
Appearance Orange powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR