4-Piperidinemethanol ≥98.3%

Synonyms: Piperidin-4-ylmethanol

02215-1 02215-2 02215-3
103534-650EA 30.77 USD
103534-650 103534-652 103534-654
4-Piperidinemethanol ≥98.3%
4-Piperidinemethanol
Formula: C₆H₁₃NO
MW: 115.18 g/mol
MDL Number: MFCD00174228
CAS Number: 6457-49-4

Order Now

Specification Test Results

Purety 98.3%
Appearance Light-Yellow crystal

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR