Thieno[3,2-d]pyrimidin-4(1H)-one ≥99.0%

Synonyms: 3H,4H-thieno[3,2-d]pyrimidin-4-one

02201-1 02201-2 02201-3
103534-644EA 30.77 USD
103534-644 103534-646 103534-648
Thieno[3,2-d]pyrimidin-4(1H)-one ≥99.0%
Thieno[3,2-d]pyrimidin-4(1H)-one
Formula: C₆H₄N₂OS
MW: 152.17 g/mol
MDL Number: MFCD00122140
CAS Number: 16234-10-9

Order Now

Specification Test Results

Purety 99.0%
Appearance Off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR