3-Bromo-2-thenoic acid ≥99.1%

Synonyms: 3-Bromo-2-thiophenecarboxylic acid, 3-Bromothiophene-2-carboxylic acid

02199-1 02199-2 02199-3
103534-638EA 30.77 USD
103534-638 103534-640 103534-642
3-Bromo-2-thenoic acid ≥99.1%
3-Bromo-2-thenoic acid
Formula: C₅H₃BrO₂S
MW: 207.05 g/mol
MDL Number: MFCD00052291
CAS Number: 7311-64-0

Order Now

Specification Test Results

Purety 99.1%
Appearance Off-white powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR