(Tetrahydrofuran-3-yl)methanamine hydrochloride ≥97.0%

Synonyms: 3-Aminomethyltetrahydrofuran hydrochloride, Oxolan-3-ylmethanamine hydrochloride, 3-(Aminomethyl)oxolane hydrochloride

02180-01-1 02180-01-2 02180-01-3
103536-050EA 138.46 USD
103536-050 103536-052 103536-054
(Tetrahydrofuran-3-yl)methanamine hydrochloride ≥97.0%
(Tetrahydrofuran-3-yl)methanamine hydrochloride
Formula: C₅H₁₂ClNO
MW: 137.61 g/mol
MDL Number: MFCD08448154
CAS Number: 184950-35-4

Order Now

Specification Test Results

Purety 97.0%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR