Tetrahydrofuran-3-ylmethanol ≥98.9%

Synonyms: Oxolan-3-ylmethanol, (Tetrahydrofuran-3-yl)methanol

02171-1 02171-2 02171-3
103536-044EA 30.77 USD
103536-044 103536-046 103536-048
Tetrahydrofuran-3-ylmethanol ≥98.9%
Tetrahydrofuran-3-ylmethanol
Formula: C₅H₁₀O₂
MW: 102.13 g/mol
MDL Number: MFCD00075198
CAS Number: 15833-61-1

Order Now

Specification Test Results

Purety 98.9%
Appearance Colorless liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR