Methyl-5-bromo-2-hydroxynicotinate ≥98.7%

Synonyms: Methyl 5-bromo-2-oxo-1,2-dihydropyridine-3-carboxylate, methyl 5-bromo-2-hydroxypyridine-3-carboxylate, Methyl 5-bromo-2-oxo-1,2-dihydro-3-pyridinecarboxylate

02140-1 02140-2 02140-3
103534-624EA 30.77 USD
103534-624 103534-626 103534-628
Methyl-5-bromo-2-hydroxynicotinate ≥98.7%
Methyl-5-bromo-2-hydroxynicotinate
Formula: C₇H₆BrNO₃
MW: 232.03 g/mol
MDL Number: MFCD03844843
CAS Number: 120034-05-1

Order Now

Specification Test Results

Purety 98.7%
Appearance Off-white powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR