2,6-Dichloronicotinic acid ≥98.4%

Synonyms: 2,6-Dichloro-3-pyridinecarboxylic acid, 2,6-Dichloropyridine-3-carboxylic acid

02098-1 02098-2 02098-3
103534-618EA 18.46 USD
103534-618 103534-620 103534-622
2,6-Dichloronicotinic acid ≥98.4%
2,6-Dichloronicotinic acid
Formula: C₆H₃Cl₂NO₂
MW: 192 g/mol
MDL Number: MFCD00075583
CAS Number: 38496-18-3

Order Now

Specification Test Results

Purety 98.4%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR