6-Azaindole ≥98.2%

02079-1 02079-2 02079-3
103534-606EA 33.85 USD
103534-606 103534-608 103534-610
6-Azaindole ≥98.2%
6-Azaindole
Formula: C₇H₆N₂
MW: 118.14 g/mol
MDL Number: MFCD01686899
CAS Number: 271-29-4

Order Now

Specification Test Results

Purety 98.2%
Appearance Off-white powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR