1H-Pyrrolo[2,3-b]pyridine-7-oxide ≥97.0%

01920-1 01920-2 01920-3
103534-594EA 123.08 USD
103534-594 103534-596 103534-598
1H-Pyrrolo[2,3-b]pyridine-7-oxide ≥97.0%
1H-Pyrrolo[2,3-b]pyridine-7-oxide
Formula: C₇H₆N₂O
MDL Number: MFCD08690076
CAS Number: 55052-24-9

Order Now

Specification Test Results

Purety 97.0%
Appearance Gray powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR