2-Amino-4-chloro-3-nitropyridine ≥97.8%

Synonyms: 4-Chloro-3-nitropyridin-2-amine

01873-1 01873-2 01873-3
103534-582EA 30.77 USD
103534-582 103534-584 103534-586
2-Amino-4-chloro-3-nitropyridine ≥97.8%
2-Amino-4-chloro-3-nitropyridine
Formula: C₅H₄ClN₃O₂
MW: 173.56 g/mol
MDL Number: MFCD04116097
CAS Number: 6980-08-1

Order Now

Specification Test Results

Purety 97.8%
Appearance Yellow powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR