Methyl-4-chloropicolinate ≥97.2%

Synonyms: Methyl 4-chloropicolinate, Methyl 4-chloropyridine-2-carboxylate

01871-1 01871-2 01871-3
103534-576EA 38.46 USD
103534-576 103534-578 103534-580
Methyl-4-chloropicolinate ≥97.2%
Methyl 4-chloropicolinate
Formula: C₇H₆ClNO₂
MW: 171.5800 g/mol
MDL Number: MFCD04116183
CAS Number: 24484-93-3

Order Now

Specification Test Results

Purety 97.2%
Appearance Yellow powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR