Methyl imidazo[1,2-a]pyridine-6-carboxylate ≥97.6%

01849-1 01849-2 01849-3
103534-570EA 69.23 USD
103534-570 103534-572 103534-574
Methyl imidazo[1,2-a]pyridine-6-carboxylate ≥97.6%
Methyl imidazo[1,2-a]pyridine-6-carboxylate
Formula: C₉H₈N₂O₂
MW: 176.1720 g/mol
MDL Number: MFCD06200984
CAS Number: 136117-69-6

Order Now

Specification Test Results

Purety 97.6%
Appearance Wheat solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR