2-Amino-4-chloropyrimidine ≥97.0%

Synonyms: 4-chloropyrimidin-2-amine

01578-1 01578-2 01578-3
103534-558EA 32.31 USD
103534-558 103534-560 103534-562
2-Amino-4-chloropyrimidine ≥97.0%
2-Amino-4-chloropyrimidine
Formula: C₄H₄ClN₃
MW: 129.55 g/mol
MDL Number: MFCD00038021
CAS Number: 3993-78-0

Order Now

Specification Test Results

Purety 97.0%
Appearance Light-Yellow powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR