2-Chloropyrimidine-5-carboxyaldehyde ≥98.9%

Synonyms: 2-Chloropyrimidine-5-carboxaldehyde, 2-Chloropyrimidine-5-carbaldehyde

01503-1 01503-2 01503-3
103534-552EA 130.77 USD
103534-552 103534-554 103534-556
2-Chloropyrimidine-5-carboxyaldehyde ≥98.9%
2-Chloropyrimidine-5-carboxyaldehyde
Formula: C₅H₃ClN₂O
MW: 142.55 g/mol
MDL Number: MFCD10696891
CAS Number: 933702-55-7

Order Now

Specification Test Results

Purety 98.9%
Appearance Off-white powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR