7-Azaindole-5-boronic acid ≥97.0%

Synonyms: (1H-Pyrrolo[2,3-b]pyridin-5-yl)boronic acid, {1H-pyrrolo[2,3-b]pyridin-5-yl}boronic acid

01423-1 01423-2 01423-3
103534-546EA 289.23 USD
103534-546 103534-548 103534-550
7-Azaindole-5-boronic acid ≥97.0%
7-Azaindole-5-boronic acid
Formula: C₇H₇BN₂O₂
MW: 161.96 g/mol
MDL Number: MFCD08234881
CAS Number: 944059-24-9

Order Now

Specification Test Results

Purety 97.0%
Appearance Off-white powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR