Methyl-3-hydroxycyclobutanecarboxylate ≥97.0%

Synonyms: Methyl 3-hydroxycyclobutanecarboxylate, Methyl 3-hydroxycyclobutane-1-carboxylate

01390-1 01390-2 01390-3
103534-540EA 61.54 USD
103534-540 103534-542 103534-544
Methyl-3-hydroxycyclobutanecarboxylate ≥97.0%
Methyl-3-hydroxycyclobutanecarboxylate
Formula: C₆H₁₀O₃
MW: 130.14 g/mol
MDL Number: MFCD00512755
CAS Number: 4934-99-0

Order Now

Specification Test Results

Purety 97.0%
Appearance Colorless liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR