Methyl-3,3-difluorocyclobutanecarboxylate ≥97.0%

Synonyms: Methyl 3,3-difluorocyclobutanecarboxylate, Methyl 3,3-difluorocyclobutane-1-carboxylate

01365-1 01365-2 01365-3
103534-534EA 46.15 USD
103534-534 103534-536 103534-538
Methyl-3,3-difluorocyclobutanecarboxylate ≥97.0%
Methyl-3,3-difluorocyclobutanecarboxylate
Formula: C₆H₈F₂O₂
MW: 150.13 g/mol
MDL Number: MFCD17015922
CAS Number: 1234616-13-7

Order Now

Specification Test Results

Purety 97.0%
Appearance Colorless liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR