tert-Butyl-3-hydroxycyclobutylcarbamate ≥97.0%

Synonyms: tert-butyl N-(3-hydroxycyclobutyl)carbamate, tert-Butyl (3-hydroxycyclobutyl)carbamate

01336-1 01336-2 01336-3
103534-528EA 61.54 USD
103534-528 103534-530 103534-532
tert-Butyl-3-hydroxycyclobutylcarbamate ≥97.0%
tert-Butyl-3-hydroxycyclobutylcarbamate
Formula: C₉H₁₇NO₃
MW: 187.24 g/mol
MDL Number: MFCD11501030
CAS Number: 154748-63-7

Order Now

Specification Test Results

Purety 97.0%
Appearance Off-white powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR