2-Azaspiro[3.3]heptane hemioxalate ≥97%

01327-01-1 01327-01-2 01327-01-3
103534-522EA 84.62 USD
103534-522 103534-524 103534-526
2-Azaspiro[3.3]heptane hemioxalate ≥97%
2-Azaspiro[3.3]heptane hemioxalate
Formula: C₁₄H₂₄N₂O₄
MW: 284.351 g/mol
MDL Number: MFCD21642066
CAS Number: 1365639-13-9

Order Now

Specification Test Results

Purety 97%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR