1-Benzyl 3-methyl azetidine-1,3-dicarboxylate ≥98.0%

Synonyms: 1-Benzyloxycarbonyl-3-azetidinecarboxylic acid methyl ester, Methyl 1-Cbz-azetidine-3-carboxylate

01230-1 01230-2 01230-3
103534-516EA 30.77 USD
103534-516 103534-518 103534-520
1-Benzyl 3-methyl azetidine-1,3-dicarboxylate ≥98.0%
1-Benzyl 3-methyl azetidine-1,3-dicarboxylate
Formula: C₁₃H₁₅NO₄
MW: 249.262 g/mol
MDL Number: MFCD11110708
CAS Number: 757239-60-4

Order Now

Specification Test Results

Purety 98.0%
Appearance Light-Yellow liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR