3-Methylazetidin-3-ol hydrochloride ≥97.0%

01226-1 01226-2 01226-3
103534-510EA 61.54 USD
103534-510 103534-512 103534-514
3-Methylazetidin-3-ol hydrochloride ≥97.0%
3-Methylazetidin-3-ol hydrochloride
Formula: C4H10ClNO
MW: 123.58 g/mol
MDL Number: MFCD11100994
CAS Number: 124668-46-8

Order Now

Specification Test Results

Purety 97.0%
Appearance White crystal

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR