1-Benzyl 3-methyl piperidine-1,3-dicarboxylate ≥98.8%

Synonyms: Methyl 1-Cbz-piperidine-3-carboxylate

01144-1 01144-2 01144-3
103534-504EA 30.77 USD
103534-504 103534-506 103534-508
1-Benzyl 3-methyl piperidine-1,3-dicarboxylate ≥98.8%
1-Benzyl 3-methyl piperidine-1,3-dicarboxylate
Formula: C₁₅H₁₉NO₄
MW: 277.316 g/mol
MDL Number: MFCD06795953
CAS Number: 174543-74-9

Order Now

Specification Test Results

Purety 98.8%
Appearance Yellow liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR