(S)-(+)-3-Amino-1-(benzyloxycarbonyl)pyrrolidine ≥98.4%

Synonyms: (S)-1-Cbz-3-aminopyrrolidine, benzyl (3S)-3-aminopyrrolidine-1-carboxylate

01090-1 01090-2 01090-3
103534-498EA 69.23 USD
103534-498 103534-500 103534-502
(S)-(+)-3-Amino-1-(benzyloxycarbonyl)pyrrolidine ≥98.4%
(S)-(+)-3-Amino-1-(benzyloxycarbonyl)pyrrolidine
Formula: C₁₂H₁₆N₂O₂
MW: 220.27 g/mol
MDL Number: MFCD06858462
CAS Number: 122536-72-5

Order Now

Specification Test Results

Purety 98.4%
Appearance Yellow oil

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR