tert-Butyl N-[(3R)-piperidin-3-ylmethyl]carbamate ≥97%

Synonyms: (R)-tert-Butyl (piperidin-3-ylmethyl)carbamate

00907-2 00907-3 00907-1
103534-494EA 176.92 USD
103534-494 103534-496 103536-042
tert-Butyl N-[(3R)-piperidin-3-ylmethyl]carbamate ≥97%
tert-Butyl N-[(3R)-piperidin-3-ylmethyl]carbamate
Formula: C₁₁H₂₂N₂O₂
MW: 214.305 g/mol
MDL Number: MFCD03093384
CAS Number: 879275-33-9
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Purety 97%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR