tert-Butyl-3-(hydroxymethyl)piperidine-1-carboxylate ≥97.0%

Synonyms: tert-Butyl 3-(hydroxymethyl)piperidine-1-carboxylate, 3-(Hydroxymethyl)piperidine, N-BOC protected

00889-1 00889-2 00889-3
103536-036EA 12.31 USD
103536-036 103536-038 103536-040
tert-Butyl-3-(hydroxymethyl)piperidine-1-carboxylate ≥97.0%
tert-Butyl-3-(hydroxymethyl)piperidine-1-carboxylate
Formula: C₁₁H₂₁NO₃
MW: 215.29 g/mol
MDL Number: MFCD03094733
CAS Number: 116574-71-1

Order Now

Specification Test Results

Purety 97.0%
Appearance Light-Yellow solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR