1-Boc-4-(hydroxymethyl)piperidine ≥97.0%

Synonyms: tert-butyl 4-(hydroxymethyl)piperidine-1-carboxylate, 1-Boc-4-piperidinemethanol, N-BOC-4-Piperidinemethanol

00888-1 00888-2 00888-3
103536-030EA 32.31 USD
103536-030 103536-032 103536-034
1-Boc-4-(hydroxymethyl)piperidine ≥97.0%
1-Boc-4-(hydroxymethyl)piperidine
Formula: C₁₁H₂₁NO₃
MW: 215.29 g/mol
MDL Number: MFCD02094488
CAS Number: 123855-51-6

Order Now

Specification Test Results

Purety 97.0%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR