(S)-1-Boc-(3-hydroxymethyl)pyrrolidine ≥97.6%

Synonyms: tert-butyl (3S)-3-(hydroxymethyl)pyrrolidine-1-carboxylate, (S)-1-Boc-(3-Hydroxymethyl)pyrrolidine

00886-1 00886-2 00886-3
103536-024EA 38.46 USD
103536-024 103536-026 103536-028
(S)-1-Boc-(3-hydroxymethyl)pyrrolidine ≥97.6%
(S)-1-Boc-(3-hydroxymethyl)pyrrolidine
Formula: C₁₀H₁₉NO₃
MW: 201.27 g/mol
MDL Number: MFCD03094726
CAS Number: 199174-24-8

Order Now

Specification Test Results

Purety 97.6%
Appearance Yellow liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR