Piperidine-4-carbonitrile hydrochloride ≥97.0%

Synonyms: 4-Cyanopiperidine hydrochloride

00851-01-1 00851-01-2 00851-01-3
103536-018EA 30.77 USD
103536-018 103536-020 103536-022
Piperidine-4-carbonitrile hydrochloride ≥97.0%
Piperidine-4-carbonitrile hydrochloride
Formula: C₆H₁₁ClN₂
MW: 146.62 g/mol
MDL Number: MFCD09037930
CAS Number: 240401-22-3

Order Now

Specification Test Results

Purety 97.0%
Appearance Yellow powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR