4-Piperidone monohydrate hydrochloride ≥97.0%

Synonyms: 4,4-Piperidinediol Hydrochloride, 4,4-Dihydroxypiperidine Hydrochloride, 4-Ketopiperidine Monohydrate Hydrochloride, 4-Piperidinone monohydrate hydrochloride, Piperidin-4-one hydrate hydrochloride, 4-Azacyclohexanone Monohydrate Hydrochloride

00847-02-1 00847-02-2 00847-02-3
103536-012EA 12.31 USD
103536-012 103536-014 103536-016
4-Piperidone monohydrate hydrochloride ≥97.0%
4-Piperidone monohydrate hydrochloride
Formula: C₅H₉NO·xHCl·yH₂O
MW: 99.13 g/mol (free base)
MDL Number: MFCD00012776
CAS Number: 40064-34-4

Order Now

Specification Test Results

Purety 97.0%
Appearance Light-Yellow powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR