5-Azaindole ≥97.0%

Synonyms: 1H-pyrrolo[3,2-c]pyridine, 1H-Pyrrolo[2,3-d]pyridine

00845-1 00845-2 00845-3
103534-492EA 30.77 USD
103534-492 103536-008 103536-010
5-Azaindole ≥97.0%
5-Azaindole
Formula: C₇H₆N₂
MW: 118.14 g/mol
MDL Number: MFCD00955936
CAS Number: 271-34-1

Order Now

Specification Test Results

Purety 97.0%
Appearance Brown solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR