4-Amino-1-methylpiperidine ≥99.2%

00820-1 00820-2 00820-3
103534-486EA 24.62 USD
103534-486 103534-488 103534-490
4-Amino-1-methylpiperidine ≥99.2%
4-Amino-1-methylpiperidine
Formula: C₆H₁₄N₂
MW: 114.1888 g/mol
MDL Number: MFCD01570549
CAS Number: 41838-46-4

Order Now

Specification Test Results

Purety 99.2%
Appearance Light-Yellow liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR