tert-Butyl N-[(3R)-piperidin-3-yl]carbamate hydrochloride ≥97%

Synonyms: (R)-3-(Boc-amino)piperidine hydrocholride

00803-01-1 00803-01-2 00803-01-3
103534-474EA 30.77 USD
103534-474 103534-476 103534-478
tert-Butyl N-[(3R)-piperidin-3-yl]carbamate hydrochloride ≥97%
tert-Butyl N-[(3R)-piperidin-3-yl]carbamate hydrochloride
Formula: C₁₀H₂₀N₂O₂·HCl
MW: 236.739 g/mol
CAS Number: 1217656-59-1

Order Now

Specification Test Results

Purety 97%
Appearance White crystal

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR