(S)-3-(Boc-amino)piperidine ≥99.9%

Synonyms: (S)-3-Boc-aminopiperidine, (S)-tert-Butyl piperidin-3-ylcarbamate, tert-butyl N-[(3S)-piperidin-3-yl]carbamate

00802-1 00802-2 00802-3
103534-468EA 30.77 USD
103534-468 103534-470 103534-472
(S)-3-(Boc-amino)piperidine ≥99.9%
(S)-3-(Boc-amino)piperidine
Formula: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
MDL Number: MFCD03093383
CAS Number: 216854-23-8

Order Now

Specification Test Results

Purety 99.9%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR