(R)-(+)-3-Hydroxypiperidine hydrochloride ≥97.0%

Synonyms: (R)-3-Piperidinol hydrochloride, (3R)-piperidin-3-ol hydrochloride, (R)-Piperidin-3-ol hydrochloride, (R)-3-Hydroxypiperidine hydrochloride

00798-01-1 00798-01-2 00798-01-3
103534-462EA 30.77 USD
103534-462 103534-464 103534-466
(R)-(+)-3-Hydroxypiperidine hydrochloride ≥97.0%
(R)-(+)-3-Hydroxypiperidine hydrochloride
Formula: C₅H₁₂ClNO
MW: 137.607 g/mol
MDL Number: MFCD00192221
CAS Number: 198976-43-1

Order Now

Specification Test Results

Purety 97.0%
Appearance White Solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR