(S)-3-Hydroxypyrrolidine hydrochloride ≥97.0%

Synonyms: (3S)-pyrrolidin-3-ol hydrochloride

00795-1-1 00795-1-2 00795-1-3
103534-456EA 38.46 USD
103534-456 103534-458 103534-460
(S)-3-Hydroxypyrrolidine hydrochloride ≥97.0%
(S)-3-Hydroxypyrrolidine hydrochloride
Formula: C₄H₁₀ClNO
MW: 123.58 g/mol
MDL Number: MFCD00272298
CAS Number: 122536-94-1

Order Now

Specification Test Results

Purety 97.0%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR