(R)-1-Boc-3-Hydroxypiperidine ≥99.8%

Synonyms: (R)-1-BOC-3-hydroxypiperidine, (R)-tert-Butyl 3-hydroxypiperidine-1-carboxylate, tert-butyl (3R)-3-hydroxypiperidine-1-carboxylate

00783-1 00783-2 00783-3
103536-002EA 30.77 USD
103536-002 103536-004 103536-006
(R)-1-Boc-3-Hydroxypiperidine ≥99.8%
(R)-1-Boc-3-hydroxypiperidine
Formula: C₁₀H₁₉NO₃
MW: 201.27 g/mol
MDL Number: MFCD04115306
CAS Number: 143900-43-0

Order Now

Specification Test Results

Purety 99.8%
Appearance White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR