1-Boc-3-hydroxypiperidine ≥97.0%

Synonyms: tert-butyl 3-hydroxypiperidine-1-carboxylate

00781-1 00781-2 00781-3
103535-996EA 32.31 USD
103535-996 103535-998 103536-000
1-Boc-3-hydroxypiperidine ≥97.0%
1-Boc-3-hydroxypiperidine
Formula: C₁₀H₁₉NO₃
MW: 201.27 g/mol
MDL Number: MFCD02093938
CAS Number: 85275-45-2

Order Now

Specification Test Results

Purety 97.0%
Appearance Light-Yellow powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR