1-Boc-4-hydroxypiperidine ≥97.0%

Synonyms: N-Boc-4-piperidinol, tert-butyl 4-hydroxypiperidine-1-carboxylate, N-BOC-4-Hydroxypiperidine, tert-Butyl 4-hydroxypiperidine-1-carboxylate

00780-1 00780-2 00780-3
103535-990EA 12.31 USD
103535-990 103535-992 103535-994
1-Boc-4-hydroxypiperidine ≥97.0%
1-Boc-4-hydroxypiperidine
Formula: C₁₀H₁₉NO₃
MW: 201.27 g/mol
MDL Number: MFCD01075174
CAS Number: 109384-19-2

Order Now

Specification Test Results

Purety 97.0%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR