(R)-(-)-1-Boc-3-hydroxypyrrolidine ≥99.5%

Synonyms: (R)-(-)-N-Boc-3-pyrrolidinol, tert-butyl (3R)-3-hydroxypyrrolidine-1-carboxylate, (R)-N-(tert-Butoxycarbonyl)-3-hydroxypyrrolidine

00779-1 00779-2 00779-3
103535-984EA 18.46 USD
103535-984 103535-986 103535-988
(R)-(-)-1-Boc-3-hydroxypyrrolidine ≥99.5%
(R)-(-)-1-Boc-3-hydroxypyrrolidine
Formula: C₉H₁₇NO₃
MW: 187.24 g/mol
MDL Number: MFCD01317838
CAS Number: 109431-87-0

Order Now

Specification Test Results

Purety 99.5%
Appearance Light-Yellow powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR