6-Hydroxypyridazine-3-carboxylic acid ≥99.9%

00733-1 00733-2 00733-3
103535-960EA 38.46 USD
103535-960 103535-962 103535-964
6-Hydroxypyridazine-3-carboxylic acid ≥99.9%
6-Hydroxypyridazine-3-carboxylic acid
Formula: C₅H₄N₂O₃
MW: 140.1000 g/mol
MDL Number: MFCD00052805
CAS Number: 37972-69-3

Order Now

Specification Test Results

Purety 99.9%
Appearance off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR