2,7-Diazaspiro[3.5]nonane-2-carboxylic acid tert-butyl ester ≥98.7%

Synonyms: tert-butyl 2,7-diazaspiro[3.5]nonane-2-carboxylate, 2-(tert-Butoxycarbonyl)-2,7-diazaspiro[3.5]nonane

00723-1 00723-2 00723-3
103535-954EA 130.77 USD
103535-954 103535-956 103535-958
2,7-Diazaspiro[3.5]nonane-2-carboxylic acid tert-butyl ester ≥98.7%
2,7-Diazaspiro[3.5]nonane-2-carboxylic acid tert-butyl ester
Formula: C₁₂H₂₂N₂O₂
MW: 226.32 g/mol
MDL Number: MFCD09834803
CAS Number: 236406-55-6

Order Now

Specification Test Results

Purety 98.7%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR