2-Oxa-7-azaspiro[3.5]nonane hemioxalate ≥97.0%

Synonyms: 2-Oxa-7-azaspiro[3.5]nonane oxalate(2:1)

00650-01-1 00650-01-2 00650-01-3
103535-930EA 230.77 USD
103535-930 103535-932 103535-934
2-Oxa-7-azaspiro[3.5]nonane hemioxalate ≥97.0%
2-Oxa-7-azaspiro[3.5]nonane hemioxalate
Formula: C₁₆H₂₈N₂O₆
MW: 344.4 g/mol
MDL Number: MFCD11035888
CAS Number: 1429056-28-9

Order Now

Specification Test Results

Purety 97.0%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR