(R)-(-)-3-Amino-1-boc-piperidine ≥97.0%

Synonyms: tert-butyl (3R)-3-aminopiperidine-1-carboxylate, (R)-1-Boc-3-Aminopiperidine

00645-1 00645-2 00645-3
103535-924EA 30.77 USD
103535-924 103535-926 103535-928
(R)-(-)-3-Amino-1-boc-piperidine ≥97.0%
(R)-(-)-3-Amino-1-Boc-piperidine
Formula: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
MDL Number: MFCD03094717
CAS Number: 188111-79-7
UN: 3259
ADR: 8,III

Order Now

Specification Test Results

Purety 97.0%
Appearance Orange liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR