Methyl-1H-indazole-4-carboxylate ≥97.0%

Synonyms: Methyl 1H-indazole-4-carboxylate

00630-1 00630-2 00630-3
103535-918EA 30.77 USD
103535-918 103535-920 103535-922
Methyl-1H-indazole-4-carboxylate ≥97.0%
Methyl 1H-indazole-4-carboxylate
Formula: C₉H₈N₂O₂
MW: 176.1700 g/mol
MDL Number: MFCD07371610
CAS Number: 192945-49-6

Order Now

Specification Test Results

Purety 97.0%
Appearance Light-Yellow powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR