1-Chloro-4-ethynylbenzene ≥99.0%

00596-1 00596-2 00596-3
103535-912EA 30.77 USD
103535-912 103535-914 103535-916
1-Chloro-4-ethynylbenzene ≥99.0%
1-Chloro-4-ethynylbenzene
Formula: C₈H₅Cl
MDL Number: MFCD00191917
CAS Number: 873-73-4

Order Now

Specification Test Results

Purety 99.0%
Appearance Yellow solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR