Methylmaleic hydrazide ≥97.0%

Synonyms: 3,6-Dihydroxy-4-methylpyridazine, 4-Methyl-3,6-pyridazinediol, 1,2-Dihydro-4-methyl-3,6-pyridazinedione, Methylmaleic Acid Hydrazide

00593-1 00593-2 00593-3
103535-906EA 84.62 USD
103535-906 103535-908 103535-910
Methylmaleic hydrazide ≥97.0%
Methylmaleic hydrazide
Formula: C₅H₆N₂O₂
MW: 126.12 g/mol
MDL Number: MFCD00067746
CAS Number: 5754-18-7

Order Now

Specification Test Results

Purety 97.0%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR