4(1H)-Pyrimidinone ≥97.0%

Synonyms: 3,4-Dihydropyrimidin-4-one, 1H-Pyrimidin-4-one, 4-Hydroxypyrimidine, Pyrimidin-4(1H)-one

00588-1 00588-2 00588-3
103535-900EA 36.92 USD
103535-900 103535-902 103535-904
4(1H)-Pyrimidinone ≥97.0%
4(1H)-Pyrimidinone
Formula: C₄H₄N₂O
MW: 96.09 g/mol
MDL Number: MFCD00006664
CAS Number: 51953-17-4

Order Now

Specification Test Results

Purety 97.0%
Appearance White crystal

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR