2,3-Dimethyl-6-nitro-1H-indazole ≥97.0%

Synonyms: 2,3-dimethyl-6-nitro-2H-indazole

00579-1 00579-2 00579-3
103535-894EA 30.77 USD
103535-894 103535-896 103535-898
2,3-Dimethyl-6-nitro-1H-indazole ≥97.0%
2,3-Dimethyl-6-nitro-1H-indazole
Formula: C₉H₁₁N₃O₂
MW: 191.1867 g/mol
MDL Number: MFCD10703326
CAS Number: 444731-73-1

Order Now

Specification Test Results

Purety 97.0%
Appearance Wheat powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR