3-Methyl-6-nitro-1H-indazole ≥97.0%

00578-1 00578-2 00578-3
103535-888EA 38.46 USD
103535-888 103535-890 103535-892
3-Methyl-6-nitro-1H-indazole ≥97.0%
3-Methyl-6-nitro-1H-indazole
Formula: C₈H₇N₃O₂
MW: 177.1600 g/mol
MDL Number: MFCD07082704
CAS Number: 6494-19-5

Order Now

Specification Test Results

Purety 97.0%
Appearance Yellow powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR